Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (S)-(+)-Carvone

Catalog No. 4357595ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-575-95ML 5 mL
43-575-925ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Warning
Catalog No. 43-575-95ML Supplier Sigma Aldrich Supplier No. 4357595ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(S)-(+)-Carvone, a monoterpene found mainly in caraway and dill seed oils, is used in cosmetic, food and pharmaceutical preparations.(S)-(+)-Carvone can be used as a starting material to synthesize: (-)-Samaderine Y, a pentacyclic quassinoid. (-)-Ambrox, a terpenoid responsible for the odor of ambergris. 3+a-Acetoxydrimenin (a sesquiterpene) via conjugated addition of potassium cyanide followed by base catalyzed Robinson annulation reaction. Thapsigargin family members such as trilobolide, nortrilobolide, and thapsivillosin F.

Specifications

CAS 2244-16-8
Density 0.96 g/mL at 25°C
Boiling Point 96°C to 98°C (10 mmHg, lit.)
Molecular Formula C10H14O
Refractive Index n20/D 1.497
Linear Formula C10H14O
MDL Number MFCD00062997
Quantity 5 mL
Synonym (+)-Carvone; (S)-5-Isopropenyl-2-methyl-2-cyclohexenone; p-Mentha-6,8-dien-2-one
Molecular Weight (g/mol) 150.22 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.