Learn More
Cayman Chemical S-CCG-1423 100ug
SDP
Catalog No. 502984275
Encompass_Preferred
A stereoisomer of CCG-1423 that more potently inhibits MRTF-A-dependent cellular events, including SRF-mediated gene expression and cell migration
Catalog No. 50-298-4275 Supplier Cayman Chemical Supplier No. 20107100 UG
A stereoisomer of CCG-1423 that more potently inhibits MRTF-A-dependent cellular events, including SRF-mediated gene expression and cell migration