Learn More
Medchemexpress LLC HY-19323A 5mg Medchemexpress, (S)-Ceralasertib CAS:1352226-87-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY19323A5MG
Encompass_Preferred
Medchemexpress, HY-19323A 5mg (S)-Ceralasertib CAS:1352226-87-9 (S)-Ceralasertib is extracted from patent WO2011154737A1, Compound II, exhibits an IC 50 of 2.578 nM [1] .(S)-Ceralasertib is a potent and selective sulfoximine morpholinopyrimidine ATR inhibitor with excellent preclinical physicochemical and pharmacokinetic (PK) characteristics.(S)-Ceralasertib is developed improving aqueous solubility and eliminates CYP3A4 time-dependent inhibition [2] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2830