Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (S)-(-)-Citronellal

Catalog No. 3737535G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
37-375-35G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 37-375-35G Supplier Sigma Aldrich Supplier No. 3737535G
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(S)-(-)-Citronellal is a monoterpenoid compound mainly found in Corymbia citriodora and Cymbopogon nardus essential oils.(S)-(-)-Citronellal may be used in the synthesis of bioactive compounds like (+)-hexahydrocannabinol, (S)-isopulegol, machaeriols A and B.

Specifications

CAS 24720-64-7
Melting Point 117°C to 121°C
Molecular Formula C10H15BrO4S -+ xH2O
Linear Formula C10H15BrO4S -+ xH2O
Refractive Index n20/D 1.446
Quantity 5 g
MDL Number MFCD00167452
Synonym (-)-Citronellal; (3S)-3,7-Dimethyl-6-octenal
Molecular Weight (g/mol) 311.19 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.