Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (S)-(+)-Citronellyl Bromide

Catalog No. 3777165G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
37-771-65G 5 g
37-771-625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Warning
Catalog No. 37-771-65G Supplier Sigma Aldrich Supplier No. 3777165G
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(S)-(+)-Citronellyl Bromide

Specifications

CAS 143615-81-0
Density 1.11 g/mL at 25°C
Boiling Point 111°C (12 mmHg, lit.)
Molecular Formula C10H19Br
Linear Formula (CH3)2C=CHCH2CH2CH(CH3)CH2CH2Br
Refractive Index n20/D 1.474
MDL Number MFCD00134459
Quantity 5 g
Synonym 8-Bromo-2,6-dimethyl-2-octene
Molecular Weight (g/mol) 219.16 g/mol
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.