Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals (S)-(+)-Clopidogrel hydrogensulfate, 98%

Catalog No. AC461030050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC461030050 5 g
AC461030010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC461030050 Supplier Thermo Scientific Chemicals Supplier No. 461030050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 120202-66-6 | C16H18ClNO6S2 | 419.89 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 120202-66-6
Molecular Formula C16H18ClNO6S2
Molecular Weight (g/mol) 419.89
MDL Number MFCD00876395,MFCD05865229,MFCD05662337
InChI Key FDEODCTUSIWGLK-RSAXXLAASA-N
Synonym clopidogrel bisulfate, clopidogrel hydrogen sulfate, iscover, plavix, clopidogrel hemisulfate, s-methyl 2-2-chlorophenyl-2-6,7-dihydrothieno 3,2-c pyridin-5 4h-yl acetate sulfate, clopidogrel sulfate, myogrel, clopidogrel sulphate, clopidogrel bisulphate
PubChem CID 115366
ChEBI CHEBI:3759
IUPAC Name methyl (2S)-2-(2-chlorophenyl)-2-(6,7-dihydro-4H-thieno[3,2-c]pyridin-5-yl)acetate;sulfuric acid
SMILES OS(O)(=O)=O.COC(=O)[C@@H](N1CCC2=C(C1)C=CS2)C1=CC=CC=C1Cl

Specifications

Loss on Drying 0.5% max.
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 5 g
Formula Weight 419.9
Percent Purity 98%
Chemical Name or Material (S)-(+)-Clopidogrel hydrogensulfate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.