Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(+)-Clopidogrel Sulfate 98.0+%, TCI America™
SDP

Catalog No. C25561G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C25561G 1 g
C25565G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C25561G Supplier TCI America Supplier No. C25561G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 120202-66-6
Molecular Formula C16H18ClNO6S2
Molecular Weight (g/mol) 419.89
MDL Number MFCD00876395,MFCD05865229,MFCD05662337
InChI Key FDEODCTUSIWGLK-RSAXXLAASA-N
Synonym clopidogrel bisulfate, clopidogrel hydrogen sulfate, iscover, plavix, clopidogrel hemisulfate, s-methyl 2-2-chlorophenyl-2-6,7-dihydrothieno 3,2-c pyridin-5 4h-yl acetate sulfate, clopidogrel sulfate, myogrel, clopidogrel sulphate, clopidogrel bisulphate
PubChem CID 115366
ChEBI CHEBI:3759
IUPAC Name methyl (2S)-2-(2-chlorophenyl)-2-{4H,5H,6H,7H-thieno[3,2-c]pyridin-5-yl}acetate; sulfuric acid
SMILES OS(O)(=O)=O.COC(=O)[C@@H](N1CCC2=C(C1)C=CS2)C1=CC=CC=C1Cl

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 419.89
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material (S)-(+)-Clopidogrel Sulfate
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.