Learn More
Cayman Chemical (S)-(+)-Clopidogrel-d3 (sulfate), Purity: ≥99% deuterated forms (d1-d3), CAS: 1217643-68-9, 5 mg
SDP
Catalog No. 502888496
Encompass_Preferred
An internal standard for the quantification of (S)-(+)-clopidogrel by GC- or LC-MS
Catalog No. 50-288-8496 Supplier Cayman Chemical Supplier No. 100106735 MG
An internal standard for the quantification of (S)-(+)-clopidogrel by GC- or LC-MS