Learn More
AdipoGen S-CR8
SDP
Supplier:  AdipoGen AGMRC0004M050
Encompass_Preferred
Chemical. CAS 1084893-56-0. Formula C24H29N7O. MW 431.5. Potent cyclin-dependent kinase CDK 1, 2, 5, and 9 inhibitor. Apoptosis inducer. Inhibits the proliferation of various cancer cell lines. Inhibits cysts formation in culture. Casein kinase 1 CK1delta/epsilon and Dyrk1A inhibitor. Down regulates expression of Mcl-1 and MYCN. Neuroprotective in experimental traumatic brain injury. Potential anti-inflammatory compound. Potential antidiabetic compound.
Catalog No. 50-256-2163