Learn More
Cayman Chemical (S)-Crizotinib, Purity: ≥98%, CAS: 1374356-45-2, 10 mg
SDP
Catalog No. 502989269
Encompass_Preferred
An MTH1 inhibitor (IC50 = 72 nM); selective for MTH1 over a panel of 456 kinases at 1 µM; induces oxidative DNA damage and apoptosis in SGC-7901 and BGC-823 cancer cells at 30 µM; induces ROS production and apoptosis in the same model; inhibits proliferation in a panel of 14 cancer cell lines expressing wild-type or various mutant forms of K-Ras or p53 (IC50s = 0.52-7.4 µM); inhibits tumor growth in an SW 480 mouse xenograft model at 25 mg/kg
Catalog No. 50-298-9269 Supplier Cayman Chemical Supplier No. 3111410 MG
An MTH1 inhibitor (IC50 = 72 nM); selective for MTH1 over a panel of 456 kinases at 1 µM; induces oxidative DNA damage and apoptosis in SGC-7901 and BGC-823 cancer cells at 30 µM; induces ROS production and apoptosis in the same model; inhibits proliferation in a panel of 14 cancer cell lines expressing wild-type or various mutant forms of K-Ras or p53 (IC50s = 0.52-7.4 µM); inhibits tumor growth in an SW 480 mouse xenograft model at 25 mg/kg