Learn More
Apexbio Technology LLC (S)-Crizotinib, 50mg. Cas: 877399-52-5;1374356-45-2 MFCD: MFCD22126092. Potent MTH1 inhibitor
SDP
Supplier:  Apexbio Technology LLC A880250
Encompass_Preferred
(S)-crizotinibthe selectively inhibited MTH1 catalytic activity with IC50 of 72 nM, while clinically used (R)-enantiomer of the drug was inactive with IC50 of 1375 nM. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5190