Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S-Curve™ Metro Rack Back-Loading Glove Dispenser

Catalog No. 19160204 Shop All S-Curve Technologies Products
Change view
Click to view available options
Color:
Clear
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-160-204 Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-160-204 Supplier S-Curve™ Supplier No. BLD2400
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Designed to fit a 24 in. deep Metro Rack the dispenser features a front access hole for product retrieval, with a rear opening for restocking product from behind the rack.

  • Specially designed to fit a 24 in. deep Metro Rack
  • Front access hole allows for easy product retrieval
  • Rear opening to restock product from behind the rack
  • Single lid with full-length hinge for durability
  • Flame treated edges for a smooth finish
  • Fit up to 3 dispensers on a 48 in. wide rack
  • Cannot be wall mounted

Specifications

Product Type Glove Dispenser
Color Clear
Dimensions (D x W x H) Interior 22 x 15 x 16 in.
Holds Gloves
Material Acrylic
No. of Compartments 1
Product Style Rear Loading
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.