Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S-Curve™ AT-100 Bulk Dispenser

Catalog No. 19034248 Shop All S-Curve Technologies Products
Change view
Click to view available options
Color:
Clear
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-034-248 Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-034-248 Supplier S-Curve™ Supplier No. AT100C
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specially designed to hold unpackaged PPE such as ambidextrous gloves, bouffant caps, beard covers, or ear plugs. Amber and gray dispensers help to protect latex gloves from ozone degradation.

  • Designed to dispense bulk PPE such as gloves, bouffant caps, or ear plugs
  • Unique access hole effectively dispenses a wide varitey of products
  • Amber and gray options help prevent ozone degradation of latex gloves
  • Can be mounted to the wall or free-standing
  • Single lid with full-length hinge for extra durability
  • Sloped top maintains HEPA flow
  • Flame treated edges to ensure a smooth finish

Specifications

Product Type Glove Dispenser
Color Clear
Dimensions (D x W x H) Interior 6 x 12 x 16 in.
Holds Gloves, Bouffant Caps, Ear Plugs
Material Acrylic
Mounting Freestanding or Wall Mount
No. of Compartments 1
Product Style Top Loading
For Use With (Application) Dispensing and storing bulk PPE
Thickness (English) 0.25 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.