Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S-Curve™ Laboratory Biohazard Shields

Catalog No. 19999948 Shop All S-Curve Technologies Products
Change view
Click to view available options
Depth (English):
6 in.
8 in.
10 in.
Width (English):
10 in.
12 in.
20 in.
Height (English):
10 in.
12 in.
16 in.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Depth (English) Width (English) Height (English)
19-999-948 6 in. 10 in. 10 in.
19-999-950 10 in. 20 in. 16 in.
19-999-949 8 in. 12 in. 12 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-999-948 Supplier S-Curve™ Supplier No. BS1010
Add to Cart
Add to Cart
This item is not returnable. View return policy

Provide a protective beta barrier

  • Constructed of 3/8 in. thick (0.95cm) clear acrylic
  • User friendly design tilts back for easy viewing of work area
  • Large base provides a secure area for working and equipment
  • Flame-treated edges for smoothness
  • Rubber pads on bottom to prevent slipping

Specifications

Depth (Metric) 15.2 cm
Width (Metric) 25.4 cm
Product Type Laboratory Beta Shield
Width (English) 10 in.
Material Acrylic
Depth (English) 6 in.
For Use With (Application) Provide a protective Beta barrier
Height (English) 10 in.
Height (Metric) 25.4 cm
Thickness (English) 0.37 in.
Thickness (Metric) 9.3 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.