Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S-Curve™ Two Compartment Sterile Packaged Gloves Dispenser

Catalog No. 19162479 Shop All S-Curve Technologies Products
Change view
Click to view available options
Color:
Clear
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-162-479 Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-162-479 Supplier S-Curve™ Supplier No. CH12
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Dispenser features two compartments, each designed to fit 5 x 6 in. glove wallets that dispense in a first-in first-out fashion.

  • Dual compartment dispenser designed for 5 x 6 in. glove wallets
  • Dispenses products in a first-in, first-out fashion
  • Sloped lid with full-length hinge to maintain HEPA flow
  • Can be mounted to the wall or free-standing
  • Flame treated edges for a smooth finish

Specifications

Product Type Glove Dispenser
Color Clear
Dimensions (D x W x H) Interior 6.5 x 11 x 16 in.
Holds Sterile Pair-Packaged Gloves
Material Acrylic
Mounting Freestanding or Wall Mount
No. of Compartments 2
Product Style Top Loading
Thickness (English) 0.25 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.