Learn More
Medchemexpress LLC (S)-Cystathionine-d4 | 61949-06-2 | 97.1% | 226.29 | 1 MG
SDP
Catalog No. 5000452069
Encompass_Preferred
(S)-Cystathionine-d4 is the deuterium labeled (S)-Cystathionine. It is for research use only and not sold to patients. It can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Deuteration has gained attention due to its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration has potential to affect pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-52069 Supplier Medchemexpress LLC Supplier No. HYW009749CS1MG
(S)-Cystathionine-d4 is the deuterium labeled (S)-Cystathionine. It is for research use only and not sold to patients. It can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Deuteration has gained attention due to its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration has potential to affect pharmacokinetic and metabolic profiles of drugs.