Learn More
Medchemexpress LLC (S)-Cystathionine-d4 | 61949-06-2 | 97.1% | 226.29 | 5 MG
SDP
Catalog No. 5000366946
Encompass_Preferred
(S)-Cystathionine-d4 is the deuterium labeled (S)-Cystathionine. It is for research use only and not sold to patients. This compound can be used as a tracer or as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Deuteration has gained attention for its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Potential to affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-003-66946 Supplier Medchemexpress LLC Supplier No. HYW009749CS5MG
(S)-Cystathionine-d4 is the deuterium labeled (S)-Cystathionine. It is for research use only and not sold to patients. This compound can be used as a tracer or as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Deuteration has gained attention for its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Potential to affect pharmacokinetic and metabolic profiles of drugs