Learn More
Cayman Chemical S-Desoxy-D2PMhydrochlorId 5mg
SDP
Catalog No. 502895383
Encompass_Preferred
Most commonly used as a chiral solvating agent for NMR analysis of chiral compounds; structurally related to desoxypipradrol, a dopamine transporter inhibitor and psychoactive stimulant
Catalog No. 50-289-5383 Supplier Cayman Chemical Supplier No. 90010955 MG
Most commonly used as a chiral solvating agent for NMR analysis of chiral compounds; structurally related to desoxypipradrol, a dopamine transporter inhibitor and psychoactive stimulant