Learn More
Medchemexpress LLC S-Diclofenac | 912758-00-0 | C23H15Cl2NO2S3 | 1 ML
SDP
Catalog No. 5000375587
Encompass_Preferred
S-Diclofenac (ACS 15) is a hybrid molecule of an H2S donor and the NSAID diclofenac. It activates the p53 signaling pathway, inhibits JNK activation, and exhibits antioxidant and anti-inflammatory activities.
  • Activates the p53 signaling pathway.
  • Inhibits the activation of JNK.
  • Exhibits antioxidant and anti-inflammatory activities.
  • Induces expression of p53 protein, leading to cell cycle arrest and apoptosis.
  • Releases hydrogen sulfide (H2S) and inhibits cyclooxygenase-mediated inflammatory responses.
  • Reverses remodeling of cardiac gap junctions, alleviates oxidative stress and inflammatory response.
Catalog No. 50-003-75587 Supplier Medchemexpress LLC Supplier No. HY1503510MM/1ML
S-Diclofenac (ACS 15) is a hybrid molecule of an H2S donor and the NSAID diclofenac. It activates the p53 signaling pathway, inhibits JNK activation, and exhibits antioxidant and anti-inflammatory activities.
  • Activates the p53 signaling pathway.
  • Inhibits the activation of JNK.
  • Exhibits antioxidant and anti-inflammatory activities.
  • Induces expression of p53 protein, leading to cell cycle arrest and apoptosis.
  • Releases hydrogen sulfide (H2S) and inhibits cyclooxygenase-mediated inflammatory responses.
  • Reverses remodeling of cardiac gap junctions, alleviates oxidative stress and inflammatory response.