Learn More
Apexbio Technology LLC (S)-(+)-Dimethindene maleate(Synonyms: (+)-Dimethindene maleate, Dexdimethindene maleate, (S)-Dimetindene maleate, (S)-(+)-Dimetindene maleate), 50mg, CAS: 136152-65-3.
SDP
Catalog No. 5000061219
Encompass_Preferred
(S)-( )-Dimethindene maleate (CAS 136152-65-3) is a small molecule antagonist exhibiting selective affinity for the muscarinic acetylcholine receptor subtype M2 while showing comparatively reduced interaction with M1 M3 and M4 subtypes Additionally it acts as an antagonist at histamine H1 receptors Due to its receptor selectivity profiles it is employed in pharmacological studies investigating autonomic regulation mechanisms cardiovascular physiology and respiratory system function
Catalog No. 50-000-61219 Supplier Apexbio Technology LLC Supplier No. B673450
(S)-( )-Dimethindene maleate (CAS 136152-65-3) is a small molecule antagonist exhibiting selective affinity for the muscarinic acetylcholine receptor subtype M2 while showing comparatively reduced interaction with M1 M3 and M4 subtypes Additionally it acts as an antagonist at histamine H1 receptors Due to its receptor selectivity profiles it is employed in pharmacological studies investigating autonomic regulation mechanisms cardiovascular physiology and respiratory system function