Learn More
Cayman Chemical S- Docosahexaenyl-1-Hyd 10mg
SDP
Catalog No. 502894948
Encompass_Preferred
A homolog of DHEA, characterized by the addition of an (S)-α-methyl group at the methylene carbon adjacent to the amide nitrogen
Catalog No. 50-289-4948 Supplier Cayman Chemical Supplier No. 900123210 MG
A homolog of DHEA, characterized by the addition of an (S)-α-methyl group at the methylene carbon adjacent to the amide nitrogen