Learn More
Medchemexpress LLC (S)-Dolaphenine hydrochloride | 135383-60-7 | 98.5% | C11H13ClN2S | 1 ML
SDP
Catalog No. 5000434409
Encompass_Preferred
(S)-Dolaphenine hydrochloride is a component used in the synthesis of Dolastatin 10, an antineoplastic agent known to inhibit tubulin polymerization. This product is intended solely for research purposes and is not sold to patients.
  • Component used in the synthesis of Dolastatin 10.
  • Precursor to an antineoplastic agent that inhibits tubulin polymerization.
  • For research use only.
  • Not for patient use.
  • Available in solid form.
  • Available in various solution concentrations.
Catalog No. 50-004-34409 Supplier Medchemexpress LLC Supplier No. HY78828A10MM/1ML
(S)-Dolaphenine hydrochloride is a component used in the synthesis of Dolastatin 10, an antineoplastic agent known to inhibit tubulin polymerization. This product is intended solely for research purposes and is not sold to patients.
  • Component used in the synthesis of Dolastatin 10.
  • Precursor to an antineoplastic agent that inhibits tubulin polymerization.
  • For research use only.
  • Not for patient use.
  • Available in solid form.
  • Available in various solution concentrations.