Learn More
Medchemexpress LLC (S)-Donepezil | 142057-80-5 | 99.8% | C24H29NO3 | 100 MG
SDP
Catalog No. 5000356324
Encompass_Preferred
(S)-Donepezil is the S-enantiomer of Donepezil, a specific and potent acetylcholinesterase (AChE) inhibitor. It is supplied as a white to off-white solid with a high purity of 99.8%. This product is intended for research use only and has not been fully validated for medical applications.
  • Specific and potent acetylcholinesterase inhibitor
  • Provided as a white to off-white solid
  • Purity of 99.8%
  • Suitable for laboratory research applications
Catalog No. 50-003-56324 Supplier Medchemexpress LLC Supplier No. HY14566A100MG
(S)-Donepezil is the S-enantiomer of Donepezil, a specific and potent acetylcholinesterase (AChE) inhibitor. It is supplied as a white to off-white solid with a high purity of 99.8%. This product is intended for research use only and has not been fully validated for medical applications.
  • Specific and potent acetylcholinesterase inhibitor
  • Provided as a white to off-white solid
  • Purity of 99.8%
  • Suitable for laboratory research applications