Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(+)-DTBM-SEGPHOS(regR) 99.0+%, TCI America™
SDP

Catalog No. D4500200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4500200MG 200 mg
D45001G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D4500200MG Supplier TCI America Supplier No. D4500200MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 210169-40-7
Molecular Formula C74H100O8P2
Molecular Weight (g/mol) 1179.554
MDL Number MFCD09753003
InChI Key ZNORAFJUESSLTM-UHFFFAOYSA-N
Synonym (S)-(+)-5,5′C-Bis[di(3,5-di-tert-butyl-4-methoxyphenyl)phosphino]-4,4′C-bi-1,3-benzodioxole, (S)-(+)-4,4′C-Bis[di(3,5-di-tert-butyl-4-methoxyphenyl)phosphino]-3,3′C-bi(1,2-methylenedioxybenzene)
PubChem CID 11194192
IUPAC Name [4-[5-bis(3,5-ditert-butyl-4-methoxyphenyl)phosphanyl-1,3-benzodioxol-4-yl]-1,3-benzodioxol-5-yl]-bis(3,5-ditert-butyl-4-methoxyphenyl)phosphane
SMILES CC(C)(C)C1=CC(=CC(=C1OC)C(C)(C)C)P(C2=C(C3=C(C=C2)OCO3)C4=C(C=CC5=C4OCO5)P(C6=CC(=C(C(=C6)C(C)(C)C)OC)C(C)(C)C)C7=CC(=C(C(=C7)C(C)(C)C)OC)C(C)(C)C)C8=CC(=C(C(=C8)C(C)(C)C)OC)C(C)(C)C

Specifications

Quantity 200 mg
Formula Weight 1179.55
Percent Purity ≥99.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material (S)-(+)-DTBM-SEGPHOS(regR)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.