Learn More
Medchemexpress LLC (S,E)-TCO-NHS ester | 1610931-22-0 | MFCD24386372 | ≥98.0% | 267.28 g/mol | C13H17NO5 | 10 MG
SDP
Catalog No. 5000286901
Encompass_Preferred
TCO-NHS ester (axial) is a trans-cyclooctene N-hydroxysuccinimide ester used as an amine-reactive building block for bioorthogonal conjugation. It enables rapid inverse electron-demand Diels-Alder (iEDDA) reactions with tetrazine partners and is suitable for labeling or modifying amine-containing small molecules and biomolecules in organic media. The reagent is supplied as a purified solid with supporting analytical documentation for research use.
  • Amine-reactive NHS ester for efficient conjugation.
  • Fast bioorthogonal iEDDA reactivity with tetrazines.
  • Suitable for labeling and bioconjugation of biomolecules.
  • High purity for reproducible experimental results.
  • Analytical documentation available (SDS, COA, H-NMR).
Catalog No. 50-002-86901 Supplier Medchemexpress LLC Supplier No. HY15169810MG
TCO-NHS ester (axial) is a trans-cyclooctene N-hydroxysuccinimide ester used as an amine-reactive building block for bioorthogonal conjugation. It enables rapid inverse electron-demand Diels-Alder (iEDDA) reactions with tetrazine partners and is suitable for labeling or modifying amine-containing small molecules and biomolecules in organic media. The reagent is supplied as a purified solid with supporting analytical documentation for research use.
  • Amine-reactive NHS ester for efficient conjugation.
  • Fast bioorthogonal iEDDA reactivity with tetrazines.
  • Suitable for labeling and bioconjugation of biomolecules.
  • High purity for reproducible experimental results.
  • Analytical documentation available (SDS, COA, H-NMR).