Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (S)-Enzaplatovir | 1323077-88-8 | 99.7% | 377.4 | C20H19N5O3 | 5 MG
SDP

Catalog No. 5000263992
Encompass_Preferred

(S)-Enzaplatovir is the S-enantiomer of Enzaplatovir, an investigational antiviral research compound active against respiratory syncytial virus (RSV). It is supplied as a solid for in vitro and in vivo studies and has reported antiviral activity (EC50 = 56 nM).

  • S-enantiomer with reported antiviral activity (EC50 = 56 nM for RSV)
  • Molecular formula C20H19N5O3; molecular weight 377.4 g·mol-1
  • High purity (99.7%) suitable for research applications
  • Soluble in DMSO (200 mg/mL); in vivo formulations achieve ≥5 mg/mL
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months)
  • Available in small research pack sizes for dosing and formulation studies

Catalog No. 50-002-63992 Supplier Medchemexpress LLC Supplier No. HY109004A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(S)-Enzaplatovir is the S-enantiomer of Enzaplatovir, an investigational antiviral research compound active against respiratory syncytial virus (RSV). It is supplied as a solid for in vitro and in vivo studies and has reported antiviral activity (EC50 = 56 nM).

  • S-enantiomer with reported antiviral activity (EC50 = 56 nM for RSV)
  • Molecular formula C20H19N5O3; molecular weight 377.4 g·mol-1
  • High purity (99.7%) suitable for research applications
  • Soluble in DMSO (200 mg/mL); in vivo formulations achieve ≥5 mg/mL
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months)
  • Available in small research pack sizes for dosing and formulation studies

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.