Learn More
Medchemexpress LLC (S)-Enzaplatovir | 1323077-88-8 | 99.7% | 377.40 | 100 MG
SDP
Catalog No. 5000346163
Encompass_Preferred
(S)-Enzaplatovir ((S)-BTA-C585) is the S-enantiomer of Enzaplatovir. It exhibits antiviral activities with an EC50 of 56 nM against respiratory syncytial viral (RSV).
  • S-enantiomer of Enzaplatovir.
  • Shows antiviral activities with an EC50 of 56 nM for respiratory syncytial viral (RSV).
Catalog No. 50-003-46163 Supplier Medchemexpress LLC Supplier No. HY109004A100MG
(S)-Enzaplatovir ((S)-BTA-C585) is the S-enantiomer of Enzaplatovir. It exhibits antiviral activities with an EC50 of 56 nM against respiratory syncytial viral (RSV).
  • S-enantiomer of Enzaplatovir.
  • Shows antiviral activities with an EC50 of 56 nM for respiratory syncytial viral (RSV).