Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (S)-(+)-Epichlorohydrin

Catalog No. 5400805G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
54-008-05G 5 g
54-008-025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 54-008-05G Supplier Sigma Aldrich Supplier No. 5400805G
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Used to synthesize inhibitors of fatty acid oxidation as potential metabolic modulators.Manufactured under license by Sterling Pharma Solutions Limited, using Jacobsen HKR technology.

Specifications

CAS 67843-74-7
Density 1.183 g/mL at 25°C
Boiling Point 92°C to 93°C360 mmHg, lit.)
Molecular Formula C3H5ClO
Refractive Index n20/D 1.438
Linear Formula C3H5ClO
MDL Number MFCD00077760
Quantity 5 g
Synonym (S)-(+)-2-(Chloromethyl)oxirane
Molecular Weight (g/mol) 92.52 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.