Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(+)-Epichlorohydrin, 98+%

Catalog No. AAL1429806
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1429806 5 g
AAL1429803 1 g
AAL1429814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1429806 Supplier Thermo Scientific Chemicals Supplier No. L1429806
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 67843-74-7 | C3H5ClO | 92.52 g/mol

(S)-(+)-Epichlorohydrin is utilized for the synthesis of macquarimicins and hydroxyisoxazolidines and (+)-cis-sylvaticin, which is a potential antitumor agent. It is used to synthesize inhibitors of fatty acid oxidation as potential metabolic modulators. It acts as a solvent for natural and synthetic resins, gums, cellulose esters and ethers, paints, varnishes, nail enamels, lacquers and cement for celluloid. It is also used as a stabilizer and also useful as a bifunctional alkylating agent with the potential to form DNA cross-links.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-(+)-Epichlorohydrin is utilized for the synthesis of macquarimicins and hydroxyisoxazolidines and (+)-cis-sylvaticin, which is a potential antitumor agent. It is used to synthesize inhibitors of fatty acid oxidation as potential metabolic modulators. It acts as a solvent for natural and synthetic resins, gums, cellulose esters and ethers, paints, varnishes, nail enamels, lacquers and cement for celluloid. It is also used as a stabilizer and also useful as a bifunctional alkylating agent with the potential to form DNA cross-links.

Solubility
Miscible with chloroform and methanol. Immiscible with water.

Notes
Hygroscopic. Store in a cool place. Incompatible with acids, bases, ammonia, amines, sodium and sodium oxides, zinc, magnesium, aluminum and its alloys and halides.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67843-74-7
Molecular Formula C3H5ClO
Molecular Weight (g/mol) 92.52
MDL Number MFCD00077760
InChI Key BRLQWZUYTZBJKN-GSVOUGTGSA-N
Synonym s-+-epichlorohydrin, s-epichlorohydrin, 2s-2-chloromethyl oxirane, s-chloromethyl oxirane, s-3-chloropropylene oxide, s-2-chloromethyl oxirane, epichlorohydrin, +, unii-scr89b4r6o, ccris 6388
PubChem CID 149428
ChEBI CHEBI:37145
IUPAC Name (2S)-2-(chloromethyl)oxirane
SMILES ClC[C@@H]1CO1

Specifications

Density 1.18
Boiling Point 116°C to 117°C
Flash Point 32°C (90°F)
Refractive Index 1.438
Quantity 5 g
UN Number UN2023
Beilstein 1420784
Solubility Information Miscible with chloroform and methanol. Immiscible with water.
Optical Rotation +35° (c=1 in Methanol)
Formula Weight 92.53
Percent Purity ≥98%
Chemical Name or Material (S)-(+)-Epichlorohydrin
Show More Show Less
Hazard Category H226-H301+H311+H331-H314-H317-H350
Hazard Statement GHS H Statement
H300-H310-H330-H350-H314-H318-H226-H317
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
May cause cancer.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P272-P280g-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: EPICHLOROHYDRIN
RTECSNumber RR0427100
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.