Learn More
Medchemexpress LLC (-)-(S)-equol | 531-95-3 | MFCD00200962 | 98.9% | 242.27 g/mol | C15H14O3 | 100 MG
SDP
Catalog No. 5000213474
Encompass_Preferred
(-)-(S)-Equol is the enantiomeric form of equol used as an analytical standard and a selective, high-affinity ligand for estrogen receptor β. It is intended for research and analytical applications, particularly receptor-binding studies and assay calibration where ERβ activity is evaluated.
  • High-affinity ERβ ligand (Ki = 0.73 nM).
  • Analytical standard suitable for research and assay calibration.
  • High purity, typically 98.9%.
  • Well-characterized chemical properties: CAS 531-95-3, C15H14O3, MW 242.27 g/mol.
  • Solid form for convenient storage and handling.
Catalog No. 50-002-13474 Supplier Medchemexpress LLC Supplier No. HY100583100MG
(-)-(S)-Equol is the enantiomeric form of equol used as an analytical standard and a selective, high-affinity ligand for estrogen receptor β. It is intended for research and analytical applications, particularly receptor-binding studies and assay calibration where ERβ activity is evaluated.
  • High-affinity ERβ ligand (Ki = 0.73 nM).
  • Analytical standard suitable for research and assay calibration.
  • High purity, typically 98.9%.
  • Well-characterized chemical properties: CAS 531-95-3, C15H14O3, MW 242.27 g/mol.
  • Solid form for convenient storage and handling.