Learn More
Cayman Chemical S-ethyl Isothiourea hydrobro
SDP
Catalog No. 502895224
Encompass_Preferred
SEIT is a potent inhibitor of NOS in vitro but lacks good in vivo efficacy due to poor cellular penetration. The Ki values are 19, 39, and 29 nM using purified human iNOS, eNOS, and nNOS, respectively.{1292,1351,2259}
Catalog No. 50-289-5224 Supplier Cayman Chemical Supplier No. 8127550 MG
SEIT is a potent inhibitor of NOS in vitro but lacks good in vivo efficacy due to poor cellular penetration. The Ki values are 19, 39, and 29 nM using purified human iNOS, eNOS, and nNOS, respectively.{1292,1351,2259}