Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S-Ethyl Trifluorothioacetate 98.0+%, TCI America™
SDP

Catalog No. T08725ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T08725ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T08725ML Supplier TCI America Supplier No. T08725ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 383-64-2
Molecular Formula C4H5F3OS
Molecular Weight (g/mol) 158.138
MDL Number MFCD00000421
InChI Key VGGUKFAVHPGNBF-UHFFFAOYSA-N
Synonym s-ethyl trifluorothioacetate, ethanethioic acid, trifluoro-, s-ethyl ester, ethyl trifluorothiolacetate, s-ethyl trifluoroethanethioate, trifluoroacetonylmercaptoethanol, acetic acid, trifluorothio-, s-ethyl ester, s-ethylthiotrifluoroacetate, trifluorothioacetic acid s-ethyl ester, ethanethioic acid, 2,2,2-trifluoro-, s-ethyl ester, 1-ethylsulfanyl-2,2,2-trifluoroethanone
PubChem CID 67844
IUPAC Name S-ethyl 2,2,2-trifluoroethanethioate
SMILES CCSC(=O)C(F)(F)F

Specifications

Color Yellow
Boiling Point 90°C
Quantity 5 mL
UN Number 1993
Formula Weight 158.14
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material S-Ethyl Trifluorothioacetate [Trifluoroacetylating Agent for Peptides Research]
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.