Learn More
Cayman Chemical S-Farnesyl Thioacetic Acid, Purity: ≥97%, CAS: 135784-48-4, 10 mg
SDP
Catalog No. 502896126
Encompass_Preferred
A competitive inhibitor of isoprenylated protein methyltransferase (also known as S-adenosylmethionine-dependent methyltransferase)
Catalog No. 50-289-6126 Supplier Cayman Chemical Supplier No. 6326010 MG
A competitive inhibitor of isoprenylated protein methyltransferase (also known as S-adenosylmethionine-dependent methyltransferase)