Learn More
Medchemexpress LLC S-Fluoxetine hydrochloride | 114247-06-2 | 99.9% | C17H19ClF3NO | 1 MG
SDP
Catalog No. 5000432683
Encompass_Preferred
S-Fluoxetine hydrochloride is an antidepressant that functions as a serotonin reuptake inhibitor (SSRI). It has a molecular weight of 345.79 and appears as a white to off-white solid. This product is designated for research use only.
  • Antidepressant
  • Serotonin reuptake inhibitor (SSRI)
  • Solid form, white to off-white in color
  • High purity of 99.9%
  • Suitable for neurological disease and neuronal signaling research
  • Supports antidepressant mechanism investigations
  • Ensures reliable experimental outcomes
  • Provides comprehensive solubility and storage guidelines
Catalog No. 50-004-32683 Supplier Medchemexpress LLC Supplier No. HYB0102B1MG
S-Fluoxetine hydrochloride is an antidepressant that functions as a serotonin reuptake inhibitor (SSRI). It has a molecular weight of 345.79 and appears as a white to off-white solid. This product is designated for research use only.
  • Antidepressant
  • Serotonin reuptake inhibitor (SSRI)
  • Solid form, white to off-white in color
  • High purity of 99.9%
  • Suitable for neurological disease and neuronal signaling research
  • Supports antidepressant mechanism investigations
  • Ensures reliable experimental outcomes
  • Provides comprehensive solubility and storage guidelines