Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (S)-Flurbiprofen | 51543-39-6 | C15H13FO2 | 100 MG
SDP

Catalog No. 5000375754
Encompass_Preferred

(S)-Flurbiprofen is an active enantiomer of Flurbiprofen, demonstrating IC50 values of 0.48 μM for COX-1 and 0.47 μM for COX-2. It effectively suppresses basal and stimulated PGE2 release in rat skin.

  • Active enantiomer of Flurbiprofen
  • COX inhibitor
  • Inhibits COX-1 with an IC50 of 0.48 μM
  • Inhibits COX-2 with an IC50 of 0.47 μM
  • Suppresses PGE2 release
  • Purity: 99.88%
  • Ee.: 99.08%
  • Appearance: Solid
  • Color: White to off-white

Catalog No. 50-003-75754 Supplier Medchemexpress LLC Supplier No. HY15123100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(S)-Flurbiprofen is an active enantiomer of Flurbiprofen, demonstrating IC50 values of 0.48 μM for COX-1 and 0.47 μM for COX-2. It effectively suppresses basal and stimulated PGE2 release in rat skin.

  • Active enantiomer of Flurbiprofen
  • COX inhibitor
  • Inhibits COX-1 with an IC50 of 0.48 μM
  • Inhibits COX-2 with an IC50 of 0.47 μM
  • Suppresses PGE2 release
  • Purity: 99.88%
  • Ee.: 99.08%
  • Appearance: Solid
  • Color: White to off-white

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.