Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (S)-Flurbiprofen | 51543-39-6 | C15H13FO2 | 500 MG
SDP

Catalog No. 5000375782
Encompass_Preferred

(S)-Flurbiprofen is an active enantiomer of Flurbiprofen, functioning as a COX inhibitor with IC50 values of 0.48 μM for COX-1 and 0.47 μM for COX-2.

  • Purity of 99.88%
  • Enantiomeric excess (ee.) of 99.08%
  • Appears as a white to off-white solid
  • Soluble in DMSO: 100 mg/mL (409.40 mM)
  • Melting point: 109-110 °C
  • Boiling point: 376.2 °C at 760 mmHg
  • Suppresses basal and stimulated PGE2 release in rat skin

Catalog No. 50-003-75782 Supplier Medchemexpress LLC Supplier No. HY15123500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(S)-Flurbiprofen is an active enantiomer of Flurbiprofen, functioning as a COX inhibitor with IC50 values of 0.48 μM for COX-1 and 0.47 μM for COX-2.

  • Purity of 99.88%
  • Enantiomeric excess (ee.) of 99.08%
  • Appears as a white to off-white solid
  • Soluble in DMSO: 100 mg/mL (409.40 mM)
  • Melting point: 109-110 °C
  • Boiling point: 376.2 °C at 760 mmHg
  • Suppresses basal and stimulated PGE2 release in rat skin

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.