Learn More
Medchemexpress LLC HY-111652 10mg Medchemexpress, S-Gboxin CAS:2101317-21-7 Purity:>98%
SDP
Catalog No. 501968936
Encompass_Preferred
Medchemexpress, HY-111652 10mg S-Gboxin CAS:2101317-21-7 S-Gboxin, a functional analogue of Gboxin, inhibits growth of mouse and human glioblastoma (GBM) with an IC50 of 470 nM. Antitumour activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8936 Supplier Medchemexpress LLC Supplier No. HY11165210MG
Add to Cart
Add to Cart
Medchemexpress, HY-111652 10mg S-Gboxin CAS:2101317-21-7 S-Gboxin, a functional analogue of Gboxin, inhibits growth of mouse and human glioblastoma (GBM) with an IC50 of 470 nM. Antitumour activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.