Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (S)-(+)-Glycidyl Benzyl Ether

Catalog No. 3635371G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
36-353-71G 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 36-353-71G Supplier Sigma Aldrich Supplier No. 3635371G
Only null left
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(S)-(+)-Glycidyl benzyl ether can be used to prepare 1-octadecyl-3-O-benzyl-sn-glycerol, which is a key intermediate for the synthesis of triglycerides containing +a-eleostearic acid.

Specifications

CAS 25526-93-6
Melting Point 176°C to 178°C
Molecular Formula C10H13FN2O4
Linear Formula C10H13FN2O4
MDL Number MFCD00056058
Quantity 25 mg
Synonym (S)-(+)-2-(Benzyloxymethyl)oxirane
Molecular Weight (g/mol) 244.22 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.