Learn More
Medchemexpress LLC HY-100742B 1mg Medchemexpress, (S)-GNE-140 CAS:2003234-64-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY100742B1MG
Encompass_Preferred
Medchemexpress, HY-100742B 1mg (S)-GNE-140 CAS:2003234-64-6 (S)-GNE-140 is the less active enantiomer of GNE-140 which can inhibit Lactate dehydrogenase A ( LDHA ). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1935