Learn More
Medchemexpress LLC (S)-Gossypol (acetic acid) | 1189561-66-7 | C32H34O10 | 100 MG
SDP
Catalog No. 5000376738
Encompass_Preferred
(S)-Gossypol is an isomer of the natural product Gossypol. It binds to the BH3-binding groove of Bcl-xL and Bcl-2 proteins with high affinity. This product is for research use only.
  • Isomer of natural product Gossypol
  • Binds to BH3-binding groove of Bcl-xL and Bcl-2 proteins
  • Acts as a Bcl-2 inhibitor
  • Available in various pack sizes
Catalog No. 50-003-76738 Supplier Medchemexpress LLC Supplier No. HY15464D100MG
(S)-Gossypol is an isomer of the natural product Gossypol. It binds to the BH3-binding groove of Bcl-xL and Bcl-2 proteins with high affinity. This product is for research use only.
  • Isomer of natural product Gossypol
  • Binds to BH3-binding groove of Bcl-xL and Bcl-2 proteins
  • Acts as a Bcl-2 inhibitor
  • Available in various pack sizes