Learn More
Apexbio Technology LLC S/GSK1349572 1051375-16-6 5mg
SDP
Catalog No. 502240329
Encompass_Preferred
S GSK1349572 (1051375-16-6) is a small-molecule inhibitor targeting HIV-1 integrase It is designed to inhibit the DNA strand transfer process interfering with viral replication by preventing the integration of viral genomic DNA into host cellular DNA S GSK1349572 exerts its biological activity primarily by binding to the integrase active site and occupying the integrase DNA interface thereby blocking DNA integration In in vitro studies using peripheral blood mononuclear cells S GSK1349572 demonstrates potent anti-HIV activity with an IC50 value of approximately 0 51 nM Based on these pharmacological properties S GSK1349572 holds research potential in antiretroviral resistance studies and HIV drug development
This item is not returnable. View return policy
Catalog No. 50-224-0329 Supplier Apexbio Technology LLC Supplier No. A40745
S GSK1349572 (1051375-16-6) is a small-molecule inhibitor targeting HIV-1 integrase It is designed to inhibit the DNA strand transfer process interfering with viral replication by preventing the integration of viral genomic DNA into host cellular DNA S GSK1349572 exerts its biological activity primarily by binding to the integrase active site and occupying the integrase DNA interface thereby blocking DNA integration In in vitro studies using peripheral blood mononuclear cells S GSK1349572 demonstrates potent anti-HIV activity with an IC50 value of approximately 0 51 nM Based on these pharmacological properties S GSK1349572 holds research potential in antiretroviral resistance studies and HIV drug development