Learn More
Vector Laboratories Sulfo S-HyNic Linker, 10 mg S-1002-010
SDP
Supplier:  Vector Laboratories S1002010
Encompass
Sulfo S-HyNic Linker (Water Soluble), 10 mg. S-HyNic heterobifunctional crosslinker is fundamental to SoluLINK bioconjugation technology. Sulfo-S-HyNic reacts with primary amines on proteins (through lysine amino acids) or amino-modified oligonucleotides or surfaces, introducing a HyNic linker that forms stable covalent conjugates with biomolecules possessing 4FB linkers. Sulfo-S-HyNic contains a Sulfo-NHS ester, which makes this linker water soluble. No DMF or other organic solvents are necessary to dissolve the linker prior to use.
Catalog No. 50-204-5740