Learn More
Apexbio Technology LLC (S)-(+)-Ibuprofen(Synonyms: Dexibuprofen, (S)-Ibuprofen, (S)-(+)-2-(4-Isobutylphenyl)propionic acid), 10mM (in 1mL DMSO), CAS: 51146-56-6.
SDP
Catalog No. 5000060608
Encompass_Preferred
(S)-( )-Ibuprofen (CAS 51146-56-6) is the pharmacologically active enantiomer of the racemic nonsteroidal anti-inflammatory drug (NSAID) ibuprofen Unlike its R(-)-counterpart the S( )-form selectively inhibits cyclooxygenase (COX) enzymes thereby suppressing prostaglandin synthesis at concentrations relevant to clinical settings Due to this stereoselectivity (S)-( )-ibuprofen is widely utilized in biomedical research investigating pain inflammation pathways COX enzyme activity and related drug-target interactions
Catalog No. 50-000-60608 Supplier Apexbio Technology LLC Supplier No. B10185.1
(S)-( )-Ibuprofen (CAS 51146-56-6) is the pharmacologically active enantiomer of the racemic nonsteroidal anti-inflammatory drug (NSAID) ibuprofen Unlike its R(-)-counterpart the S( )-form selectively inhibits cyclooxygenase (COX) enzymes thereby suppressing prostaglandin synthesis at concentrations relevant to clinical settings Due to this stereoselectivity (S)-( )-ibuprofen is widely utilized in biomedical research investigating pain inflammation pathways COX enzyme activity and related drug-target interactions