Learn More
Medchemexpress LLC (S)-(+)-Ibuprofen-d3 | 1329643-44-8 | MFCD30748182 | 100.0% | 209.30 g/mol | C13H15D3O2 | 10 MG
SDP
Catalog No. 5000299251
Encompass_Preferred
(S)-(+)-Ibuprofen-d3 is the S-enantiomer of ibuprofen labeled with three deuterium atoms, supplied as a solid analytical standard for use in analytical, pharmacokinetic, and tracer studies. It is provided as a high-purity, stable powder intended for research applications only.
  • High isotopic labeling suitable for use as an internal standard.
  • High purity (reported 99.96%) for analytical accuracy.
  • White to off-white solid form for easy handling and storage.
  • Stable under recommended storage conditions (powder: -20°C).
  • Available in small quantity sizes for analytical workflows.
Catalog No. 50-002-99251 Supplier Medchemexpress LLC Supplier No. HY78131AS10MG
Add to Cart
Add to Cart
(S)-(+)-Ibuprofen-d3 is the S-enantiomer of ibuprofen labeled with three deuterium atoms, supplied as a solid analytical standard for use in analytical, pharmacokinetic, and tracer studies. It is provided as a high-purity, stable powder intended for research applications only.
  • High isotopic labeling suitable for use as an internal standard.
  • High purity (reported 99.96%) for analytical accuracy.
  • White to off-white solid form for easy handling and storage.
  • Stable under recommended storage conditions (powder: -20°C).
  • Available in small quantity sizes for analytical workflows.