Learn More
Medchemexpress LLC HY-111540B 10mg Medchemexpress, (S)-IDO1-IN-5 CAS:2166616-76-6 Purity:>98%
SDP
Catalog No. 501968908
Encompass_Preferred
Medchemexpress, HY-111540B 10mg (S)-IDO1-IN-5 CAS:2166616-76-6 (S)-IDO1-IN-5 (Example 1B) is an active S-isomer of IDO1-IN-5. (S)-IDO1-IN-5 binds to IDOL with an IC50 value less than 1.5 µΜ. IDO1-IN-5 is a potent, selective and brain penetrated inhibitor of Indoleamine 2,3-Dioxygenase 1 (IDO1) activity, binds to apo-IDO1 lacking heme rather than mature heme-bound IDO1. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8908 Supplier Medchemexpress LLC Supplier No. HY111540B10MG
Medchemexpress, HY-111540B 10mg (S)-IDO1-IN-5 CAS:2166616-76-6 (S)-IDO1-IN-5 (Example 1B) is an active S-isomer of IDO1-IN-5. (S)-IDO1-IN-5 binds to IDOL with an IC50 value less than 1.5 µΜ. IDO1-IN-5 is a potent, selective and brain penetrated inhibitor of Indoleamine 2,3-Dioxygenase 1 (IDO1) activity, binds to apo-IDO1 lacking heme rather than mature heme-bound IDO1. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.