Learn More
Apexbio Technology LLC S-INDOXIMOD-200MG
SDP
Catalog No. 5000569650
Encompass_Preferred
(S)-Indoximod (CAS No 21339-55-9) is a small molecule inhibitor primarily used in immuno-oncology research It acts by targeting the indoleamine 2 3-dioxygenase (IDO) pathway which is pivotal in tumor immune evasion mechanisms By inhibiting IDO (S)-Indoximod helps in modulating immune responses potentially enhancing anti-tumor immunity Its in vitro activity is generally observed in the nanomolar to low micromolar range Common applications include studies involving various cell lines such as human cancer models and animal studies to explore therapeutic effects in vivo It is also utilized in drug screening platforms to evaluate potential combinatorial cancer therapies Experimental concentrations typically depend on the specific research design allowing for flexibility in diverse investigative contexts
Catalog No. 50-005-69650 Supplier Apexbio Technology LLC Supplier No. C6586200
(S)-Indoximod (CAS No 21339-55-9) is a small molecule inhibitor primarily used in immuno-oncology research It acts by targeting the indoleamine 2 3-dioxygenase (IDO) pathway which is pivotal in tumor immune evasion mechanisms By inhibiting IDO (S)-Indoximod helps in modulating immune responses potentially enhancing anti-tumor immunity Its in vitro activity is generally observed in the nanomolar to low micromolar range Common applications include studies involving various cell lines such as human cancer models and animal studies to explore therapeutic effects in vivo It is also utilized in drug screening platforms to evaluate potential combinatorial cancer therapies Experimental concentrations typically depend on the specific research design allowing for flexibility in diverse investigative contexts