Learn More
Medchemexpress LLC (S)-Indoximod-d3 | 1801851-87-5 | 221.27 | 5 MG
SDP
Catalog No. 5000415466
Encompass_Preferred
(S)-Indoximod-d3 is the deuterium labeled (S)-Indoximod. (S)-Indoximod (1-Methyl-L-tryptophan) is an inhibitor of indoleamine 2,3-dioxygenase (IDO). (S)-Indoximod can be used for the research of cancer.
  • Application: Metabolism-protein/nucleotide metabolism.
Catalog No. 50-004-15466 Supplier Medchemexpress LLC Supplier No. HYN0707S5MG
(S)-Indoximod-d3 is the deuterium labeled (S)-Indoximod. (S)-Indoximod (1-Methyl-L-tryptophan) is an inhibitor of indoleamine 2,3-dioxygenase (IDO). (S)-Indoximod can be used for the research of cancer.
  • Application: Metabolism-protein/nucleotide metabolism.