Learn More
Medchemexpress LLC (S)-Indoximod-d3 | 1801851-87-5 | MFCD11983540 | 99.0% | 221.27 g/mol | C12H11D3N2O2 | 10 MG
SDP
Catalog No. 5000296209
Encompass_Preferred
(S)-Indoximod-d3 is the deuterium-labeled isotopologue of (S)-indoximod (1-methyl-L-tryptophan), supplied as a research reference standard for biochemical and pharmacokinetic studies of the indoleamine-2,3-dioxygenase (IDO) pathway. It is intended for use as an internal standard, tracer, or analytical reference in mass spectrometry and related assays.
  • Deuterium-labeled internal standard for quantitative mass spectrometry
  • High isotopic and chemical purity (99.0%) suitable for tracer and PK studies
  • Compact 10 mg quantity for analytical applications
  • Useful in biochemical studies of IDO pathway and immunometabolism
  • CAS number provided for unambiguous chemical identification
Catalog No. 50-002-96209 Supplier Medchemexpress LLC Supplier No. HYN0707S10MG
(S)-Indoximod-d3 is the deuterium-labeled isotopologue of (S)-indoximod (1-methyl-L-tryptophan), supplied as a research reference standard for biochemical and pharmacokinetic studies of the indoleamine-2,3-dioxygenase (IDO) pathway. It is intended for use as an internal standard, tracer, or analytical reference in mass spectrometry and related assays.
  • Deuterium-labeled internal standard for quantitative mass spectrometry
  • High isotopic and chemical purity (99.0%) suitable for tracer and PK studies
  • Compact 10 mg quantity for analytical applications
  • Useful in biochemical studies of IDO pathway and immunometabolism
  • CAS number provided for unambiguous chemical identification