Learn More
Cayman Chemical S-isopropyl Isothiourea (hydrobromide), Purity: ≥98%, CAS: 4269-97-0, 50 mg
SDP
Catalog No. 502895223
Encompass_Preferred
IPTU is a potent inhibitor of NOS in vitro but may lack good in vivo efficacy due to poor cellular penetration. The Ki values are 9.8, 22, and 37 nM using purified human iNOS, eNOS, and nNOS, respectively.{1292}
Catalog No. 50-289-5223 Supplier Cayman Chemical Supplier No. 8129050 MG
IPTU is a potent inhibitor of NOS in vitro but may lack good in vivo efficacy due to poor cellular penetration. The Ki values are 9.8, 22, and 37 nM using purified human iNOS, eNOS, and nNOS, respectively.{1292}