Learn More
Apexbio Technology LLC S-Isopropylisothiourea hydrobromide 4269-97-0 500mg
SDP
Catalog No. 5000092066
Encompass_Preferred
S-Isopropylisothiourea hydrobromide is a small-molecule inhibitor targeting nitric oxide synthase (NOS) It is designed to inhibit NOS activity thereby reducing nitric oxide (NO) production and modulating NO-mediated biological processes S-Isopropylisothiourea hydrobromide exerts its biological activity primarily through selective NOS inhibition Based on these pharmacological properties S-Isopropylisothiourea hydrobromide holds research potential in investigating NO-related processes particularly in pathophysiological contexts such as hemorrhagic shock
Catalog No. 50-000-92066 Supplier Apexbio Technology LLC Supplier No. B6494500
S-Isopropylisothiourea hydrobromide is a small-molecule inhibitor targeting nitric oxide synthase (NOS) It is designed to inhibit NOS activity thereby reducing nitric oxide (NO) production and modulating NO-mediated biological processes S-Isopropylisothiourea hydrobromide exerts its biological activity primarily through selective NOS inhibition Based on these pharmacological properties S-Isopropylisothiourea hydrobromide holds research potential in investigating NO-related processes particularly in pathophysiological contexts such as hemorrhagic shock